Assay Details
| Gene Symbol: Dgk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103749.2 | NP_001097219.1 | 16 - 17 | 2656 | 62 | |
| NM_001103750.3 | NP_001097220.1 | 15 - 16 | 2347 | 62 | ||
| NM_001273863.1 | NP_001260792.1 | 15 - 16 | 2323 | 62 | ||
| NM_001316408.1 | NP_001303337.1 | 15 - 16 | 2323 | 62 | ||
| NM_078930.4 | NP_523654.2 | 5 - 6 | 1261 | 62 | ||
| NM_165568.3 | NP_724619.2 | 17 - 18 | 2713 | 62 | ||
| NM_206053.2 | NP_995775.2 | 4 - 5 | 1129 | 62 | ||
| GenBank mRNA | BT003323.1 | - | 5 - 6 | 969 | 62 | |
More Information
Set Membership: |
|

Dgk
Genomic Map