Assay Details
| Gene Symbol: Sep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143704.3 | NP_651961.1 | 2 | 1522 | 121 | |
| NM_165578.3 | NP_724632.1 | 1 | 1582 | 121 | ||
| GenBank mRNA | AF167578.1 | - | 2 | 1480 | 121 | |
| AY051754.1 | - | 1 | 1530 | 121 | ||
| BT099601.1 | - | 2 | 1482 | 121 | ||
More Information
Set Membership: |
|

Sep5
Genomic Map