Assay Details
| Gene Symbol: dap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057600.6 | NP_476948.1 | 2 - 3 | 1184 | 67 | |
| GenBank mRNA | BT125118.1 | - | 2 - 3 | 860 | 67 | |
| U68477.1 | - | 2 - 3 | 1196 | 67 | ||
More Information
Set Membership: |
|

dap
Genomic Map