Assay Details
| Gene Symbol: caz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272673.1 | NP_001259602.1 | 2 - 3 | 277 | 55 | |
| NM_078641.4 | NP_523365.2 | 2 - 3 | 277 | 55 | ||
| GenBank mRNA | BT004875.1 | - | 2 - 3 | 241 | 55 | |
More Information
Set Membership: |
|

caz
Genomic Map