Assay Details
| Gene Symbol: caz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272672.1 | NP_001259601.1 | 2 - 3 | 250 | 65 | |
| NM_078641.4 | NP_523365.2 | 3 - 4 | 382 | 65 | ||
| NM_167493.2 | NP_727946.2 | 3 - 4 | 337 | 65 | ||
| GenBank mRNA | BT004875.1 | - | 3 - 4 | 346 | 65 | |
More Information
Set Membership: |
|

caz
Genomic Map