Assay Details
| Gene Symbol: caz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272672.1 | NP_001259601.1 | 5 - 6 | 898 | 131 | |
| NM_001272673.1 | NP_001259602.1 | 5 - 6 | 995 | 131 | ||
| NM_078641.4 | NP_523365.2 | 6 - 7 | 1030 | 131 | ||
| NM_167493.2 | NP_727946.2 | 6 - 7 | 985 | 131 | ||
| GenBank mRNA | BT004875.1 | - | 6 - 7 | 994 | 131 | |
More Information
Set Membership: |
|

caz
Genomic Map