Assay Details
| Gene Symbol: Arp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103531.2 | NP_001097001.1 | 6 - 7 | 1351 | 63 | |
| NM_001272692.1 | NP_001259621.1 | 6 - 7 | 1588 | 63 | ||
| NM_078648.4 | NP_523372.2 | 5 - 6 | 1590 | 63 | ||
| GenBank mRNA | AY061256.1 | - | 5 - 6 | 1590 | 63 | |
| BT126051.1 | - | 6 - 7 | 1331 | 63 | ||
More Information
Set Membership: |
|

Arp2
Genomic Map