Assay Details
| Gene Symbol: mRpL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078655.4 | NP_523379.2 | 2 - 3 | 618 | 62 | |
| GenBank mRNA | AY070969.1 | - | 2 - 3 | 597 | 62 | |
More Information
Set Membership: |
|

mRpL22
Genomic Map