Assay Details
| Gene Symbol: Cdc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258833.3 | NP_001245762.1 | 2 - 3 | 448 | 71 | |
| NM_001258834.3 | NP_001245763.1 | 2 - 3 | 375 | 71 | ||
| NM_078690.4 | NP_523414.1 | 2 - 3 | 448 | 71 | ||
| GenBank mRNA | AY119570.1 | - | 1 - 2 | 481 | 71 | |
| U11824.1 | - | 2 - 3 | 449 | 71 | ||
More Information
Set Membership: |
|

Cdc42
Genomic Map