Assay Details
| Gene Symbol: AP-1-2beta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078691.3 | NP_523415.1 | 2 - 3 | 1531 | 74 | |
| GenBank mRNA | BT016054.1 | - | 2 - 3 | 1514 | 74 | |
| X75910.1 | - | 1510 | 74 | |||
More Information
Set Membership: |
|

AP-1-2beta
Genomic Map