Assay Details
| Gene Symbol: slgA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258867.2 | NP_001245796.1 | 6 - 7 | 1048 | 78 | |
| NM_078709.4 | NP_523433.2 | 6 - 7 | 1405 | 78 | ||
| NM_167752.3 | NP_728423.1 | 6 - 7 | 1405 | 78 | ||
More Information
Set Membership: |
|

slgA
Genomic Map