Assay Details
| Gene Symbol: Swip-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299150.1 | NP_001286079.1 | 1 - 2 | 615 | 74 | |
| NM_136081.3 | NP_609925.1 | 1 - 2 | 615 | 74 | ||
| GenBank mRNA | AB081653.1 | - | 1 - 2 | 517 | 74 | |
| AY094963.1 | - | 1 - 2 | 588 | 74 | ||
More Information
Set Membership: |
|

Swip-1
Genomic Map