Assay Details
| Gene Symbol: Dgk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103749.2 | NP_001097219.1 | 5 - 6 | 941 | 112 | |
| NM_165568.3 | NP_724619.2 | 5 - 6 | 941 | 112 | ||
More Information
Set Membership: |
|

Dgk
Genomic Map