Assay Details
| Gene Symbol: hts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259491.2 | NP_001246420.1 | 11 - 12 | 2317 | 106 | |
| NM_001299683.1 | NP_001286612.1 | 9 - 10 | 2119 | 106 | ||
| NM_166347.3 | NP_725886.2 | 11 - 12 | 2317 | 106 | ||
| NM_166348.3 | NP_725887.2 | 11 - 12 | 2307 | 106 | ||
| NM_166349.3 | NP_725888.2 | 10 - 11 | 2553 | 106 | ||
| GenBank mRNA | AF151705.1 | - | 11 - 12 | 2279 | 106 | |
More Information
Set Membership: |
|

hts
Genomic Map