Assay Details
| Gene Symbol: hts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202048.3 | NP_001188977.2 | 11 - 12 | 2216 | 80 | |
| NM_001259491.2 | NP_001246420.1 | 12 - 13 | 2400 | 80 | ||
| NM_001259492.2 | NP_001246421.1 | 12 - 13 | 2265 | 80 | ||
| NM_001299683.1 | NP_001286612.1 | 10 - 11 | 2202 | 80 | ||
More Information
Set Membership: |
|

hts
Genomic Map