Assay Details
| Gene Symbol: hts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103907.3 | NP_001097377.2 | 5 - 6 | 930 | 64 | |
| NM_001169732.2 | NP_001163203.1 | 2 - 3 | 686 | 64 | ||
| NM_001169733.2 | NP_001163204.1 | 4 - 5 | 726 | 64 | ||
| NM_001169734.2 | NP_001163205.1 | 5 - 6 | 920 | 64 | ||
| NM_001169735.2 | NP_001163206.1 | 4 - 5 | 1166 | 64 | ||
| NM_001202048.3 | NP_001188977.2 | 3 - 4 | 677 | 64 | ||
| NM_001259491.2 | NP_001246420.1 | 5 - 6 | 930 | 64 | ||
| NM_001259492.2 | NP_001246421.1 | 4 - 5 | 726 | 64 | ||
| NM_001299683.1 | NP_001286612.1 | 3 - 4 | 732 | 64 | ||
| NM_001382109.1 | NP_001369098.1 | 5 - 6 | 930 | 64 | ||
| NM_057529.4 | NP_476877.1 | 5 - 6 | 930 | 64 | ||
| NM_166347.3 | NP_725886.2 | 5 - 6 | 930 | 64 | ||
| NM_166348.3 | NP_725887.2 | 5 - 6 | 920 | 64 | ||
| NM_166349.3 | NP_725888.2 | 4 - 5 | 1166 | 64 | ||
| NM_166350.2 | NP_725889.1 | 5 - 6 | 930 | 64 | ||
| NM_166351.2 | NP_725890.1 | 5 - 6 | 930 | 64 | ||
| GenBank mRNA | AF151705.1 | - | 5 - 6 | 892 | 64 | |
| AF151706.1 | - | 4 - 5 | 700 | 64 | ||
| AF151707.1 | - | 2 - 3 | 683 | 64 | ||
More Information
Set Membership: |
|

hts
Genomic Map