Assay Details
| Gene Symbol: hts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103907.3 | NP_001097377.2 | 9 - 10 | 2175 | 75 | |
| NM_001169734.2 | NP_001163205.1 | 9 - 10 | 2165 | 75 | ||
| NM_001169735.2 | NP_001163206.1 | 8 - 9 | 2411 | 75 | ||
| NM_001202048.3 | NP_001188977.2 | 7 - 8 | 1922 | 75 | ||
| NM_001259491.2 | NP_001246420.1 | 9 - 10 | 2175 | 75 | ||
| NM_001259492.2 | NP_001246421.1 | 8 - 9 | 1971 | 75 | ||
| NM_001299683.1 | NP_001286612.1 | 7 - 8 | 1977 | 75 | ||
| NM_001382109.1 | NP_001369098.1 | 9 - 10 | 2175 | 75 | ||
| NM_057529.4 | NP_476877.1 | 9 - 10 | 2175 | 75 | ||
| NM_166347.3 | NP_725886.2 | 9 - 10 | 2175 | 75 | ||
| NM_166348.3 | NP_725887.2 | 9 - 10 | 2165 | 75 | ||
| NM_166349.3 | NP_725888.2 | 8 - 9 | 2411 | 75 | ||
| NM_166350.2 | NP_725889.1 | 9 - 10 | 2175 | 75 | ||
| GenBank mRNA | AF151705.1 | - | 9 - 10 | 2137 | 75 | |
More Information
Set Membership: |
|

hts
Genomic Map