Assay Details
| Gene Symbol: Pu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079077.4 | NP_523801.2 | 3 - 4 | 857 | 63 | |
| NM_166430.3 | NP_726037.1 | 3 - 4 | 738 | 63 | ||
| NM_166431.4 | NP_726038.1 | 3 - 4 | 906 | 63 | ||
| GenBank mRNA | AY051890.1 | - | 3 - 4 | 889 | 63 | |
| BT125829.1 | - | 3 - 4 | 857 | 63 | ||
More Information
Set Membership: |
|

Pu
Genomic Map