Assay Details
| Gene Symbol: Syb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144161.2 | NP_001137633.1 | 3 - 4 | 526 | 62 | |
| NM_001299349.1 | NP_001286278.1 | 3 - 4 | 419 | 62 | ||
| NM_078956.4 | NP_523680.1 | 3 - 4 | 457 | 62 | ||
| GenBank mRNA | BT009942.1 | - | 3 - 4 | 442 | 62 | |
| BT132995.1 | - | 3 - 4 | 419 | 62 | ||
| KX531346.1 | - | 2 - 3 | 311 | 62 | ||
| X76200.1 | - | 455 | 62 | |||
More Information
Set Membership: |
|

Syb
Genomic Map