Assay Details
| Gene Symbol: mIF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299413.1 | NP_001286342.1 | 1 - 2 | 408 | 91 | |
| NM_001363928.1 | NP_001350856.1 | 1 - 2 | 408 | 91 | ||
| NM_136915.4 | NP_610759.2 | 1 - 2 | 408 | 91 | ||
| GenBank mRNA | BT022487.1 | - | 1 - 2 | 334 | 91 | |
More Information
Set Membership: |
|

mIF3
Genomic Map