Assay Details
| Gene Symbol: sei | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299885.1 | NP_001286814.1 | 11 - 12 | 2394 | 68 | |
| NM_057365.4 | NP_476713.1 | 10 - 11 | 1913 | 68 | ||
| NM_166653.2 | NP_726408.1 | 10 - 11 | 1949 | 68 | ||
| GenBank mRNA | AY058350.1 | - | 10 - 11 | 1899 | 68 | |
| U36925.1 | - | 10 - 11 | 1884 | 68 | ||
More Information
Set Membership: |
|

sei
Genomic Map