Assay Details
| Gene Symbol: sei | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299885.1 | NP_001286814.1 | 3 - 4 | 1405 | 92 | |
| NM_057365.4 | NP_476713.1 | 2 - 3 | 924 | 92 | ||
| NM_166653.2 | NP_726408.1 | 2 - 3 | 960 | 92 | ||
| GenBank mRNA | AY058350.1 | - | 2 - 3 | 910 | 92 | |
More Information
Set Membership: |
|

sei
Genomic Map