Assay Details
| Gene Symbol: eEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138034.3 | NP_611878.1 | 2 - 3 | 252 | 85 | |
| NM_166654.2 | NP_726411.1 | 2 - 3 | 303 | 85 | ||
| GenBank mRNA | AY071396.1 | - | 2 - 3 | 281 | 85 | |
| KX531679.1 | - | 1 - 2 | 196 | 85 | ||
More Information
Set Membership: |
|

eEF5
Genomic Map