Assay Details
| Gene Symbol: Dg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079032.4 | NP_523756.1 | 7 - 8 | 1372 | 62 | |
| GenBank mRNA | AF461101.1 | - | 6 - 7 | 724 | 62 | |
| AY069076.1 | - | 7 - 8 | 1345 | 62 | ||
More Information
Set Membership: |
|

Dg
Genomic Map