Assay Details
| Gene Symbol: Dg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079032.4 | NP_523756.1 | 8 - 9 | 1619 | 86 | |
| NM_176183.3 | NP_788363.1 | 9 - 10 | 2414 | 86 | ||
| GenBank mRNA | AF461101.1 | - | 7 - 8 | 971 | 86 | |
| AY069076.1 | - | 8 - 9 | 1592 | 86 | ||
More Information
Set Membership: |
|

Dg
Genomic Map