Assay Details
| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079105.4 | NP_523829.2 | 1 - 2 | 370 | 60 | |
| GenBank mRNA | BT083409.1 | - | 1 - 2 | 366 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg3| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079105.4 | NP_523829.2 | 1 - 2 | 370 | 60 | |
| GenBank mRNA | BT083409.1 | - | 1 - 2 | 366 | 60 | |
Set Membership: |
|