Assay Details
| Gene Symbol: bgcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299858.1 | NP_001286787.1 | 2 - 3 | 2075 | 77 | |
| NM_079108.3 | NP_523832.2 | 2 - 3 | 2090 | 77 | ||
| GenBank mRNA | AF255662.1 | - | 2 - 3 | 1960 | 77 | |
More Information
Set Membership: |
|

bgcn
Genomic Map