Assay Details
| Gene Symbol: zip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014552.3 | NP_001014552.1 | 1 - 2 | 153 | 97 | |
| NM_001259587.2 | NP_001246516.1 | 1 - 2 | 153 | 97 | ||
| NM_079136.4 | NP_523860.2 | 1 - 2 | 153 | 97 | ||
More Information
Set Membership: |
|

zip
Genomic Map