Assay Details
| Gene Symbol: RfC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079191.3 | NP_523915.1 | 1 - 2 | 218 | 71 | |
| GenBank mRNA | AY094829.1 | - | 1 - 2 | 199 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RfC4| Gene Symbol: RfC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079191.3 | NP_523915.1 | 1 - 2 | 218 | 71 | |
| GenBank mRNA | AY094829.1 | - | 1 - 2 | 199 | 71 | |
Set Membership: |
|