Assay Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079219.3 | NP_523943.1 | 2 - 3 | 1073 | 55 | |
| NM_168151.3 | NP_729118.1 | 3 - 4 | 852 | 55 | ||
| NM_168152.3 | NP_729119.1 | 2 - 3 | 793 | 55 | ||
| GenBank mRNA | AY070540.1 | - | 2 - 3 | 853 | 55 | |
More Information
Set Membership: |
|

Rcc1
Genomic Map