Assay Details
| Gene Symbol: Cp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079255.2 | NP_523979.1 | 2 | 349 | 58 | |
| GenBank mRNA | BT031038.1 | - | 2 | 348 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cp15| Gene Symbol: Cp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079255.2 | NP_523979.1 | 2 | 349 | 58 | |
| GenBank mRNA | BT031038.1 | - | 2 | 348 | 58 | |
Set Membership: |
|