Assay Details
| Gene Symbol: Nrx-IV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079310.3 | NP_524034.2 | 11 - 12 | 4000 | 67 | |
| NM_168491.3 | NP_729787.2 | 11 - 12 | 4000 | 67 | ||
| GenBank mRNA | BT120004.1 | - | 11 - 12 | 3967 | 67 | |
More Information
Set Membership: |
|

Nrx-IV
Genomic Map