Assay Details
| Gene Symbol: Ten-m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104191.3 | NP_001097661.2 | 7 - 8 | 4160 | 60 | |
| NM_001275282.1 | NP_001262211.1 | 7 - 8 | 4160 | 60 | ||
| NM_079491.3 | NP_524215.2 | 5 - 6 | 2489 | 60 | ||
More Information
Set Membership: |
|

Ten-m
Genomic Map