Assay Details
| Gene Symbol: Rcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274034.2 | NP_001260963.1 | 1 - 2 | 650 | 94 | |
| NM_001274036.2 | NP_001260965.1 | 1 - 2 | 650 | 94 | ||
| NM_001299475.1 | NP_001286404.1 | 1 - 2 | 650 | 94 | ||
| NM_137083.4 | NP_610927.3 | 1 - 2 | 650 | 94 | ||
| GenBank mRNA | AY069502.1 | - | 1 - 2 | 625 | 94 | |
More Information
Set Membership: |
|

Rcd1
Genomic Map