Assay Details
| Gene Symbol: unc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299521.1 | NP_001286450.1 | 3 - 4 | 1553 | 69 | |
| NM_137189.3 | NP_611033.2 | 2 - 3 | 1351 | 69 | ||
| GenBank mRNA | AY058501.1 | - | 2 - 3 | 1325 | 69 | |
More Information
Set Membership: |
|

unc-5
Genomic Map