Assay Details
| Gene Symbol: unc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299521.1 | NP_001286450.1 | 6 - 7 | 2066 | 81 | |
| NM_137189.3 | NP_611033.2 | 5 - 6 | 1864 | 81 | ||
| GenBank mRNA | AY058501.1 | - | 5 - 6 | 1838 | 81 | |
More Information
Set Membership: |
|

unc-5
Genomic Map