Assay Details
| Gene Symbol: unc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299521.1 | NP_001286450.1 | 8 - 9 | 2506 | 65 | |
| NM_137189.3 | NP_611033.2 | 7 - 8 | 2304 | 65 | ||
| GenBank mRNA | AY058501.1 | - | 7 - 8 | 2278 | 65 | |
More Information
Set Membership: |
|

unc-5
Genomic Map