Assay Details
| Gene Symbol: ovo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169202.2 | NP_001162673.1 | 1 - 2 | 1263 | 76 | |
| NM_167026.4 | NP_726971.1 | 1 - 2 | 1263 | 76 | ||
| GenBank mRNA | AY094838.1 | - | 1 - 2 | 1239 | 76 | |
More Information
Set Membership: |
|

ovo
Genomic Map