Assay Details
| Gene Symbol: ovo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038742.3 | NP_001033831.1 | 3 - 4 | 4069 | 74 | |
| NM_001169202.2 | NP_001162673.1 | 3 - 4 | 4727 | 74 | ||
| NM_080338.5 | NP_525077.2 | 4 - 5 | 3910 | 74 | ||
| NM_167026.4 | NP_726971.1 | 3 - 4 | 4727 | 74 | ||
| NM_167027.4 | NP_726972.1 | 3 - 4 | 3482 | 74 | ||
| GenBank mRNA | AY094838.1 | - | 3 - 4 | 4703 | 74 | |
| AY119649.1 | - | 4 - 5 | 3910 | 74 | ||
More Information
Set Membership: |
|

ovo
Genomic Map