Assay Details
| Gene Symbol: apolpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272156.1 | NP_001259085.1 | 6 - 7 | 9757 | 70 | |
| NM_001297791.1 | NP_001284720.1 | 6 - 7 | 9808 | 70 | ||
| NM_001297792.1 | NP_001284721.1 | 6 - 7 | 9882 | 70 | ||
| NM_079895.3 | NP_524634.2 | 6 - 7 | 9757 | 70 | ||
More Information
Set Membership: |
|

apolpp
Genomic Map