Assay Details
| Gene Symbol: veil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299604.1 | NP_001286533.1 | 5 - 6 | 881 | 67 | |
| NM_137373.3 | NP_611217.1 | 4 - 5 | 682 | 67 | ||
| GenBank mRNA | AY069716.1 | - | 4 - 5 | 705 | 67 | |
More Information
Set Membership: |
|

veil
Genomic Map