Assay Details
| Gene Symbol: veil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299604.1 | NP_001286533.1 | 7 - 8 | 1477 | 83 | |
| NM_137373.3 | NP_611217.1 | 6 - 7 | 1278 | 83 | ||
| GenBank mRNA | AY069716.1 | - | 6 - 7 | 1301 | 83 | |
More Information
Set Membership: |
|

veil
Genomic Map