Assay Details
| Gene Symbol: Tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259488.2 | NP_001246417.1 | 2 - 3 | 2012 | 68 | |
| NM_001274157.1 | NP_001261086.1 | 2 - 3 | 1956 | 68 | ||
| NM_137564.3 | NP_611408.2 | 2 - 3 | 1956 | 68 | ||
| GenBank mRNA | BT010287.1 | - | 2 - 3 | 1915 | 68 | |
| BT100309.1 | - | 2 - 3 | 2096 | 68 | ||
More Information
Set Membership: |
|

Tab2
Genomic Map