Assay Details
| Gene Symbol: Mpcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299699.1 | NP_001286628.1 | 3 - 4 | 1068 | 61 | |
| NM_001299700.1 | NP_001286629.1 | 3 - 4 | 994 | 61 | ||
| NM_137624.2 | NP_611468.1 | 3 - 4 | 1114 | 61 | ||
| GenBank mRNA | AY121712.1 | - | 3 - 4 | 1112 | 61 | |
| BT120211.1 | - | 3 - 4 | 1068 | 61 | ||
More Information
Set Membership: |
|

Mpcp1
Genomic Map