Assay Details
| Gene Symbol: mnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167581.3 | NP_728104.1 | 8 - 9 | 3658 | 54 | |
| NM_170659.2 | NP_728107.2 | 7 - 8 | 2379 | 54 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mnb| Gene Symbol: mnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167581.3 | NP_728104.1 | 8 - 9 | 3658 | 54 | |
| NM_170659.2 | NP_728107.2 | 7 - 8 | 2379 | 54 | ||
Set Membership: |
|