Assay Details
| Gene Symbol: CG1801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167739.3 | NP_728408.3 | 9 - 10 | 3903 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG1801| Gene Symbol: CG1801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167739.3 | NP_728408.3 | 9 - 10 | 3903 | 70 | |
Set Membership: |
|