Assay Details
| Gene Symbol: Cdc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130524.5 | NP_569880.1 | 1 - 2 | 1144 | 88 | |
| GenBank mRNA | AF081540.1 | - | 1 - 2 | 1092 | 88 | |
| AY051861.1 | - | 1 - 2 | 1102 | 88 | ||
More Information
Set Membership: |
|

Cdc45
Genomic Map