Assay Details
| Gene Symbol: inc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297857.1 | NP_001284786.1 | 3 - 4 | 2011 | 89 | |
| NM_001297858.1 | NP_001284787.1 | 1 - 2 | 579 | 89 | ||
| NM_130570.3 | NP_569926.2 | 1 - 2 | 579 | 89 | ||
| GenBank mRNA | BT126052.1 | - | 1 - 2 | 546 | 89 | |
More Information
Set Membership: |
|

inc
Genomic Map