Assay Details
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130593.4 | NP_569949.3 | 1 | 1109 | 48 | |
| NM_166910.2 | NP_726771.1 | 2 | 1295 | 48 | ||
| GenBank mRNA | AY118757.1 | - | 2 | 1295 | 48 | |
More Information
Set Membership: |
|

Pex5
Genomic Map