Assay Details
| Gene Symbol: Coa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272218.1 | NP_001259147.1 | 1 - 2 | 92 | 55 | |
| NM_130600.2 | NP_569956.1 | 1 - 2 | 341 | 55 | ||
More Information
Set Membership: |
|

Coa8
Genomic Map